missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for column chromatography,ultra pure, 40-63μm, 40A

Catalog No. AC360031000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC360031000 100 g
AC360035000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360031000 Supplier Thermo Scientific Chemicals Supplier No. 360031000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7631-86-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

Chemical Identifiers

CAS 7631-86-9

Specifications

Particle Size 40 to 60 μm
Pore Size 40 Å
Packaging Plastic Bottle
Quantity 100 g
Product Type Silica Gel
Material Silica Gel
For Use With (Application) Column Chromatography

RUO – Research Use Only