missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for chromatography, 0.200-0.500 mm, 40 A

CAS: 7631-86-9 | O2Si | 60.08 g/mol

$305.68 - $305.68

Chemical Identifiers

CAS 7631-86-9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC241670010
SDS View Documents
Thermo Scientific Chemicals
241670010
1 kg
Each for $305.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

Specifications

Chemical Identifiers

7631-86-9

Specifications

40 Å
1 kg
Silica Gel
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
Plastic Bottle
Silica Gel
O2Si
7631-86-9
O=[Si]=O
60.08
Videos
SDS
Documents

Documents

RUO – Research Use Only