missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for chromatography, 0.035-0.070 mm, 60 A

For column chromatography. | CAS: 7631-86-9

Supplier:  Thermo Scientific Chemicals 240360050

 View more versions of this product

Catalog No. AC240360050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
Specifications

Specifications

60 Å
5 kg
Silica Gel
7631-86-9
Plastic Drum
Silica Gel
Column Chromatography

Chemical Identifiers

7631-86-9
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only