missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Silica gel, for chromatography, 0.035-0.070 mm, 60 A
For column chromatography. | CAS: 7631-86-9
Supplier: Thermo Scientific Chemicals 240360010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographySpecifications
| 60 Å | |
| 1 kg | |
| Silica Gel | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione |
| Plastic Bottle | |
| Silica Gel | |
| O2Si | |
| 7631-86-9 | |
| O=[Si]=O | |
| 60.08 |
Chemical Identifiers
| 7631-86-9 |
RUO – Research Use Only