missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Silica gel, for chromatography, 0.200-0.500 mm, 60 A
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications:
For column chromatography
Specifications
Specifications
| Pore Size | 60 Å |
| Packaging | Plastic Bottle |
| Quantity | 1 kg |
| Type | Silica Gel |
| Material | Silica Gel |
| Molecular Formula | O2Si |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| SMILES | O=[Si]=O |
| IUPAC Name | silanedione |
| Show More |
RUO – Research Use Only