missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for chromatography, 0.075-0.250 mm, 150 A

CAS: 7631-86-9 | O2Si | 60.08 g/mol

$215.81 - $215.81

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC419282500
SDS View Documents
Thermo Scientific Chemicals
419282500
250 g Glass bottle
Each for $215.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
24261
O=[Si]=O

Specifications

White
7631-86-9
Nominal particle size: >60 mesh: <7.5%, <200 mesh: <10.0%
250 g
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Solubility in water: insoluble.
O=[Si]=O
60.08
CHEBI:30563
Silica gel, For column chromatography
Very Fine Granular Powder
Glass bottle
275 to 375 m2/g (BET)
O2Si
15, 8632
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
60.08
Videos
SDS
Documents

Documents

RUO – Research Use Only