missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica gel, for chromatography, 0.075-0.250 mm, 150 A
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
Specifications
Specifications
| Color | White |
| Physical Form | Very Fine Granular Powder |
| Packaging | Plastic Drum |
| Particle Size | Nominal particle size: >60 mesh: <7.5%, <200 mesh: <10.0% |
| Specific Surface Area | 275 to 375 m2/g (BET) |
| Quantity | 5 kg |
| Merck Index | 15, 8632 |
| Solubility Information | Solubility in water: insoluble. |
| IUPAC Name | silanedione |
| Formula Weight | 60.08 |
| Show More |
RUO – Research Use Only