missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for chromatography, 0.035-0.070 mm, 90 A

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Supplier:  Thermo Scientific Chemicals 241650010

 View more versions of this product

Catalog No. AC241650010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
Specifications

Specifications

Silica gel
7631-86-9
1 kg
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Silicon dioxide
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08
For chromatography, 0.035-0.070mm, 90Å
Silica Gel
O2Si
Plastic Bottle
Solubility in water: insoluble.
O=[Si]=O
60.08
CHEBI:30563

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Silicon dioxide
CHEBI:30563
O=[Si]=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only