missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica gel, for chromatography, 0.030-0.200 mm, 60 A
CAS: 7631-86-9 | O2Si | 60.08 g/mol
$430.85 - $430.85
Chemical Identifiers
| CAS | 7631-86-9 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| Synonym | Silicon dioxide |
| PubChem CID | 24261 |
| ChEBI | CHEBI:30563 |
| SMILES | O=[Si]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC419290010
|
Thermo Scientific Chemicals
419290010 |
1 kg | Glass Bottle |
Each for $430.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| 24261 | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| Silicon dioxide | |
| CHEBI:30563 |
Specifications
| Powder | |
| 7631-86-9 | |
| 1 kg | |
| 6.0% max. (160°C) | |
| 15, 8632 | |
| 30 Micron D(10% min.), 250 Micron D(90% Max.) | |
| 0.8 to 1.0mL/g (N2) | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| 24261 | |
| 60.08 | |
| Silica gel, 0.060-0.200mm, 60 A |
| White | |
| Silica Gel | |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| Glass Bottle | |
| Silicon dioxide | |
| Solubility in water: insoluble. | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:30563 | |
| 500 to 600 m2/g |
RUO – Research Use Only