missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Silica Gel C8-Reversed Phase, Fully Endcapped
Description
Reversed-phase silica gels are generally suitable to retain catalysts by hydrophobic interactions.
Specifications
Specifications
| Pore Size | 90 Å |
| Grade | For column chromatography |
| Quantity | 50 g |
| MDL Number | MFCD00148339 |