Learn More
MilliporeSigma™ Silica gel 60 PF254 for preparative thin layer chromatography, MilliporeSigma™
Catalog No. M1077471000
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1077471000 1 kg
M1077472500 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1077471000 Supplier MilliporeSigma™ Supplier No. 1.07747.1000
Only null left
Silica gel 60 PF254 for preparative thin layer chromatography. CAS No. 7631-86-9, EC Number 231-545-4.
Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40μm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
Specifications
Grade 60 PF254
Quantity 1 kg
Material Silica Gel