Learn More
Silica gel 60, with fluorescent indicator, 0.060-0.2mm (70-230 mesh)
Dry column chromatography without applied pressure (gravity pulled) | CAS: 7631-86-9 | O2Si | 60.08 g/mol
$46.78 - $2026.58
Chemical Identifiers
| CAS | 7631-86-9 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| PubChem CID | 24261 |
| ChEBI | CHEBI:30563 |
| IUPAC Name | dioxosilane |
| SMILES | O=[Si]=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4275718
|
Thermo Scientific Chemicals
04275718 |
50 g |
Each for $46.78
|
|
|||||
|
AA4275730
|
Thermo Scientific Chemicals
04275730 |
250 g |
Each for $143.34
|
|
|||||
|
AA42757A1
|
Thermo Scientific Chemicals
042757A1 |
1 kg |
Each for $428.97
|
|
|||||
|
AA42757A7
|
Thermo Scientific Chemicals
042757A7 |
5 kg |
Each for $2,026.58
|
|
|||||
Description
Silica gel 60 is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used in dry column chromatography without applied pressure (gravity pulled).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| CHEBI:30563 | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| 24261 | |
| dioxosilane |
Specifications
| -70+230 Mesh Powder,S.A. 500-600m | |
| Silica Gel | |
| O2Si | |
| 70-230 Mesh | |
| 0.060 to 0.2mm | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| dioxosilane | |
| 24261 | |
| 500 to 600m2/g |
| 7631-86-9 | |
| 50 g | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| Odorless | |
| Insoluble in water. | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:30563 | |
| Silica gel 60, With fluorescent indicator |
Safety and Handling
GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 231-545-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only