missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, with fluorescent indicator, 0.060-0.2mm (70-230 mesh)

Catalog No. AA42757A7
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42757A7 5 kg
AA4275718 50 g
AA4275730 250 g
AA42757A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA42757A7 Supplier Thermo Scientific Chemicals Supplier No. 042757A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dry column chromatography without applied pressure (gravity pulled) | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica gel 60 is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used in dry column chromatography without applied pressure (gravity pulled).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form -70+230 Mesh Powder,S.A. 500-600m
Material Silica Gel
Quantity 5 kg
Mesh Size 70-230 Mesh
Odor Odorless
Particle Size 0.060 to 0.2mm
Solubility Information Insoluble in water.
Surface Area 500 to 600m2/g
Chemical Name or Material Silica gel 60, With fluorescent indicator
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only