missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel, 60%

Catalog No. 111012536
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012536 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012536 Supplier MilliporeSigma™ Supelco™ Supplier No. 899431KGF

Enriched with ∼0.1% Ca to protect acid labile protection groups. For column chromatrography.

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.

Specifications

Pore Size 60 Å
Grade For column chromatography
Quantity 1 kg
Surface Area (Metric) 500 m2/g (approx.)
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8