missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, 60 to 200 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5391670
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2003891 500 g Wide Mouth Amber Glass Bottle
2003892 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2003891 Supplier Avantor Supplier No. 341401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

    Suitable For Column Chromatography

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Material Silica Gel
Quantity 500 g
Mesh Size 60 to 200 Mesh
Packaging Wide Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Silica Gel