missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, 60 to 200 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Numéro de catalogue. 2003891
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
2003891 500 g Wide Mouth Amber Glass Bottle
2003892 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 2003891 Fournisseur Avantor Code fournisseur 341401

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

High quality chemicals for laboratory and specialized industrial use

    Suitable For Column Chromatography

Identifiants chimiques

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Spécifications

Material Silica Gel
Quantity 500 g
Mesh Size 60 to 200 Mesh
Packaging Wide Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Silica Gel