missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.062-0.105mm (150-230 mesh)

Catalog No. p-7052178
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Catalog No. Quantity
AA4272718 50 g
AA4272730 250 g
AA42727A1 1 kg
AA42727A7 5 kg
4 options

Catalog No. AA4272718

Supplier: Thermo Scientific Chemicals 4272718

Only null left

Column chromatography without applied pressure (gravity pulled) | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form -150+230 Mesh,S.A. 500-600m
Material Silica Gel
Quantity 50 g
Mesh Size 150-230 Mesh
Odor Odorless
Particle Size 0.062 to 0.105mm
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Surface Area 500 to 600m2/g
Chemical Name or Material Silica gel 60
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only