missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.060-0.2mm (70-230 mesh)

Catalog No. AAL140020E
Change view
Click to view available options
Quantity:
1000 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL140020E 2500 g
AAL140020B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL140020E Supplier Thermo Scientific Chemicals Supplier No. L140020E

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and removing impurities. It is used as stationary base in column chromatography.

Solubility
Soluble in hydrofluoric acid. Insoluble in water.

Notes
Keep the container tightly sealed. Store in cool, dry conditions in a well sealed container. Incompatible with hydrogen fluoride.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 2500 g
Mesh Size 70 to 230 mesh
Odor Odorless
Merck Index 14,8493
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Chemical Name or Material Silica gel 60, 0.060 to 0.2mm
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
RTECSNumber VV7565000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only