missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Supplier:  Thermo Scientific Chemicals 042570A1

 View more versions of this product

Catalog No. AA42570A1


Only null left
Explore available promotions

Description

Description

Column chromatographySilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Powder, -230+400 Mesh,S.A. 500-600m
7631-86-9
Odorless
O2Si
White
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
Silica gel 60
230-400 Mesh
99.5% (metals basis)
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
1 kg
Soluble in hydrofluoric acid. Insoluble in water.
O=[Si]=O
60.08
CHEBI:30563

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H373: May cause damage to organs through prolonged or repeated exposure.

EINECSNumber : 231-545-4

RTECSNumber : VV7565000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only