missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.036-0.071mm (215-400 mesh)

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Supplier:  Thermo Scientific Chemicals L140040E

 View more versions of this product

Catalog No. AAL140040E


Only null left
Explore available promotions

Description

Description

Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

Solubility
Soluble in hydrofluoric acid. Insoluble in water.

Notes
Keep the container tightly sealed. Store in cool, dry conditions in a well sealed container. Incompatible with hydrogen fluoride.
Specifications

Specifications

Silica gel 60
7631-86-9
2500 g
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
14,8493
Soluble in hydrofluoric acid. Insoluble in water.
O=[Si]=O
60.08
CHEBI:30563
0.036 to 0.071mm
Silica Gel
O2Si
215 to 400 mesh
Odorless
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
24261
dioxosilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 231-545-4

RTECSNumber : VV7565000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only