missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel

Catalog No. 111016081
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016081 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016081 Supplier MilliporeSigma™ Supelco™ Supplier No. S839450EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Desiccant

Silica gel, highly porous textured matrix are generally used for synthesis of adsorbents. They are corpuscular in structure. Silica surface possess a layer of hydroxyl groups and adsorbed water molecules, which can be removed by drying at high temperature or even by treatment with suitable reagents. Silica gel surface plays important role in catalysis, chemical separation and microelectronic fabrications.

Specifications

Pore Size 60 Å
Grade Reagent Grade
Packaging Porous 1 x 23/4 in. packet
Quantity 5 g
Surface Area (Metric) 500 m2/g (approx.)
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8