missing translation for 'onlineSavingsMsg'
Learn More

Silica 63-100μm, 60Å, EcoChrom™, MP Biomedicals™

For Chromatography; Chemical Synthesis

$8335.06 - $8335.06

Specifications

Content And Storage Store at Room Temperature.
Type Silica
Physical Form Powder
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Density 2g/cm3 at 20°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN04660
View Documents
MP Biomedicals Inc
0204660
25 kg
Each for $8,335.06
Only null left
 
Description

Description

Specifications

Specifications

Store at Room Temperature.
Powder
2g/cm3 at 20°C
Silica
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
7631-86-9
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'