missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silica 36-200μm, 60Å, EcoChrom™, MP Biomedicals™
For Chromatography; Chemical Synthesis
Manufacturer: Mp Biomedicals 0204662
Specifications
Silica | |
O2Si | |
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite | |
O=[Si]=O | |
60.083 | |
CHEBI:30563 | |
Powder | |
500-600m2/g | |
White |
7631-86-9 | |
MFCD00011232 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08 | |
2g/cm3 at 20°C | |
60Å | |
500g |
Chemical Identifiers
7631-86-9 | |
60.083 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232 | |
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite | |
CHEBI:30563 | |
O=[Si]=O |
Safety and Handling
Recommended Storage : Store at Room Temperature.