missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaSphere™ Spherical Silica Gel, 5 μm, 100 Å

Catalog No. p-7238050
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
01184342 5 g
1 options

Catalog No. 01184342

Supplier: SiliCycle™ S10005EA5G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly polar sorbent for the purification of non-polar and non-ionic compounds.

SiliaSphere- Bulk Analytical Spherical Silica Gels

SiliaSphere, the right choice for:

  • Chromatographic performance
  • Loading capacity
  • Reproducibility
  • Chemical & physical stability

With pore diameters ranging from 60 to 1000 Å and particle sizes from 1.8 to 25 μm, we SiliCycle™ products to meet all your separation requirements. This is one of the most reliable portfolios of spherical silica gels for high pressure chromatography.

SiliaSphere are ideal for both analytical and preparative chromatography, from laboratory to pilot-plant processes and production scales. Furthermore, the excellent properties of SiliaSphere make them the packing of choice for high performance liquid chromatography (HPLC), Supercritical Fluid Chromatography (SFC) and simulated moving bed (SMB) applications.

Features and Benefits of SiliaSphere™ Spherical Silica Gels:

  • High purity silica gels (Consistency, reliability & reproducibility)
  • Perfect spherical shape, free of any cavities or cracks (Ease of column packing and high resolution)
  • Exceptional narrow particle size distribution (Optimal separation and resolution)
  • Strong mechanical stability (Low back-pressure without surface abrasion)
  • Same well controlled processes for all SiliaSphere (Easy scalability)

SiliaSphere as a Silica Matrix

SiliCycle has a strong know-how and expertise in silica gel manufacturing. To support the increasing demand on spherical silicas, SiliCycle developed an optimized and highly controlled large-scale production process for all of their SiliaSphere products, without decreasing the quality of the silica.

Particle shape, pore & particle size distributions, silica gel purity and surface properties, all have their influence on chromatographic performance. Therefore, in order to develop the most efficient process, all these parameters need to be evaluated and optimized to ensure batch-to-batch reproducibility.

SiliaSphere is manufactured from an organic form of silicon (alkoxydes). This ensures very low metal content as the starting material is purified by distillation. Deionized water is used to hydrolyze the silicon alkoxydes. Careful monitoring and control of the parameters that induce precipitation provide spherical silica gels with the desired characteristics.

SiliaSphere products are characterized by a very low metal content and exceptional stability. Furthermore, SiliCycle™'s manufacturing process ensures quality and reproducibility in pore size, surface area, particle size and morphology for all SiliaSphere products.

Specifications

Particle Size 5 μm
Pore Size 100 Å
Stationary Phase Silica
Technique Liquid Chromatography
Packaging Bulk
Quantity 5 g
Sorbent Bare Silica (Silicon Dioxyde, SiO2)
Content And Storage Keep dry
Product Line SiliaSphere
Type Bulk Spherical Silica Gel
Material Silica Gel
For Use With (Application) Chromatography, purification
CAS 7631-86-9
Endcapped No
Particle Shape Spherical
Show More Show Less

Keep dry