missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SiliCycle™ SiliaMetS™ Metal Scavengers TAAcONa nec, 40 to 63 μm, 60 Å
Our TAAcONa Metal Scavenger is a version of EDTA in its sodium salt form powerful for trapping Ca, Cd, Cs, Cu, Fe, Ir, La, Li, Mg, Ni, Os, Rh, Sc and Sn.
Specifications
| Chemical Name or Material | Bulk Functionalized Irregular Silica Gel |
|---|---|
| Physical Form | Irregular |
| Packaging | HDPE Bottle |
| Product Line | SiliaMetS |
| Density | 0.712 g/mL |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01184016
|
SiliCycle™
R69230B100G |
100 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
SiliaMetS™ TAAcOH and TAAcONa are supported versions of EDTA in their acid and sodium salt forms. These two products are effective Metal Scavengers for Ca, Mg, Li, Ir, Cs, Os, Sn, Pd, Ni and Cu. SiliaMetS ™ TAAcONa is useful for metals in higher oxidation states (≥2) (Best for: Ca, Cd, Cs, Cu, Fe, Ir, La, Li, Mg, Ni, Os, Rh, Sc and Sn; also good for: Cr, Pd, Ru and Zn)
- Only need to filter out the scavenger once it has trapped the residual metal or organic impurity
- Great variety of formats for all purifications scales, from laboratory to process to plant scales
- Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
| Bulk Functionalized Irregular Silica Gel | |
| HDPE Bottle | |
| 0.712 g/mL |
| Irregular | |
| SiliaMetS |