missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaMetS™ Metal Scavengers Imidazole (IMI), 40 to 63 μm, 60 Å

Catalog No. p-7238214
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01184040 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01184040 Supplier SiliCycle™ Supplier No. R79230B100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Our Imidazole is a powerful scavenger for the removal of Cd, Co, Cu, Fe, Ir, Li, Mg, Ni, Os, W and Zn, as well as Cr, Pd and Rh.

SiliaMetS™ Imidazole is a versatile Metal Scavenger for a variety of metals, but it is best for Cd, Co, Cu, Fe, Ir, Li, Mg, Ni, Os, W and Zn. It is also good for Cr, Pd and Rh. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our Imidazole offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed

Specifications

Chemical Name or Material Bulk Functionalized Irregular Silica Gel
Physical Form Irregular
Quantity 100 g
Packaging HDPE Bottle
Product Line SiliaMetS
Density 0.681 g/mL