missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Quick Connect Dust Mop Frame, 48 in.

Catalog No. 01182368
Change view
Click to view available options
Size:
48 in. (123 cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01182368 48 in. (123 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01182368 Supplier Contec™ Supplier No. MGMP0010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mop frame intended to use with the MicroGenesis Disposable Microfiber Mop Pad, which is a 100% synthetic, single-use cleaning solution designed for ISO Class 8 environments and above. It effectively captures fine particles and large debris, providing superior contaminant removal in both wet and dry applications.

Ideal for controlled environments, cleanrooms, and general facility cleaning, the MicroGenesis Disposable Microfiber Mop Pad offers a reliable, hygienic, and effective solution for maintaining pristine floors.

  • Compatible with common disinfectants like quasts, peroxides and bleach
  • Simplifies quality control, inventory and space management, all without crosscontamination risk
  • Provides efficient, effective removal of surface contamination
  • Use MicroGenesis Mop Pads with MicroGenesis Hygienic Backer Plate and appropriate hardware

Specifications

Product Type Mop Frame
Color Gray
Sterility Non-sterile
Autoclavable Not Autoclavable
Size 48 in. (123 cm)
For Use With (Equipment) For use with MGMP0006 mop pads; QuickConnect Handle.
Color-coded No
Material Aluminum