missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Pentafluorophenyl (PFP), 40 to 63 μm, 60 Å

Catalog No. 01182357
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01182357 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01182357 Supplier SiliCycle™ Supplier No. R67530B5G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SiliaBond™ Pentafluorophenyl (PFP), 40 - 63 μm, 60 Å is a phase with a new selectivity approach with aromatic ring interactions for the purification of conjugated compounds (isomers).

  • Sorbent primarily used in the separation of molecules bearing fluorine atoms but may also be used in the separation of non-fluorous compounds such as Taxol™ and its derivatives.
  • Si-PFP has a higher selectivity for aromatics containing molecules compared to the other fluorinated sorbents due to its phenyl ring.

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase PFP (Pentafluorophenyl)
Technique Column Chromatography
Quantity 5 g
Sorbent PFP (Pentafluorophenyl)
Surface Area (Metric) 500 m2/g
Content And Storage Keep dry.
Product Line SiliaBond
Type Bulk Functionalized Irregular Silica Gel
Material Silica Gel
For Use With (Equipment) Glass Gravity Columns
For Use With (Application) Purification of polar and aromatic compounds, Separation of isomers
Endcapped Yes
Carbon Load 11%
Show More Show Less