missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Diol nec, 40 to 63 μm, 60 Å

Catalog No. 01182253
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01182253 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01182253 Supplier SiliCycle™ Supplier No. R35030B5G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Our Diol is a multifunctional gel, a chromatographic phase and an organic scavenger for trapping boronic acids.

SiliaBond™ Diol acts as a Chromatographic Phase and an Organic Scavenger for Electrophiles and Lewis Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our Diol offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • For difficult separation of low to medium polarity samples
  • Can be used in HILIC mode
  • For mono and polysaccharides separation
  • Covalent and Ionic Bonding for Boronic Acids
  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Diol nec
Technique Column Chromatography
Grade Irregular Silica Gel
Quantity 5 g
Sorbent Diol (Dihydroxypropyl) nec
Surface Area (Metric) 500 m2/g
Content And Storage Keep Dry
Product Line SiliaBond
Type Bulk Functionalized Irregular Silica Gel
Material Silica Gel
For Use With (Equipment) Bulk or Glass Gravity Columns
For Use With (Application) Purification of slightly polar compounds and HILIC mode
Physical Form Irregular
Density 0.687 g/mL
Endcapped No
Packing Material HDPE Bottle
Particle Shape Irregular
Show More Show Less