missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Amine (WAX) nec, 40 to 63 μm, 60 Å

Catalog No. 01182301
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01182301 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01182301 Supplier SiliCycle™ Supplier No. R52130B5G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This gel amine is a multifunctional chromatographic phase, powerful for trapping Cd, Cr, Pd, Pt, Rh and Ru and also very efficient toward Co, Cu, Fe, Hg, Pb, W and Zn.

Also known for their electrophile scavenging efficiencies and base reagent qualities, SiliaMetS™ Amine, Diamine and Triamine have also proven to be very useful for the scavenging of Pd, Pt, Cr, W and Zn. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Gel is great for a variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Acts as a Metal Scavenger (best for: Cd, Cr, Pd, Pt, Rh and Ru; also good for: Co, Cu, Fe, Hg, Pb, W and Zn)
  • Acts as an Organic Scavenger for Electrophiles and Acids (Covalent Binding for Acyl Chlorides, Aldehydes, Anhydrides, Chloroformates, Isocyanates, Ketones and
    Sulfonyl Chlorides; Ionic Binding and Catch and Release for Acids and Acidic Phenols)
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Amine (WAX) nec
Technique Column Chromatography
Grade Irregular Silica Gel
Quantity 5 g
Sorbent Amine (3-Aminopropyl, NH2, WAX) nec
Surface Area (Metric) 500 m2/g
Content And Storage Keep Cool (<8°C) and Dry
Product Line SiliaBond
Product Type Bulk Functionalized Irregular Silica Gel
Material Silica Gel
For Use With (Equipment) Bulk or Glass Gravity Columns
For Use With (Application) Bulk Scavenger in Post-reaction Mixture or Purification of Strong Acidic Molecules
Physical Form Irregular
Density 0.700 g/mL
Endcapped No
Packing Material HDPE Bottle
Particle Shape Irregular
Show More Show Less