Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule.
| Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Description | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Description | Price | Quantity & Availability | |||
|
03150514
|
Thermo Scientific™
290MA201 |
Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits | Pk. of 10 Ferrules | 290 MA201 |
N/A
|
|
|||
|
03050250
|
Thermo Scientific™
290MA202 |
All SilTite Kits | Pk. of 10 Ferrules | 290 MA202 |
N/A
|
|
|||
|
03050251
|
Thermo Scientific™
290MA203 |
All SilTite Kits | Pk. of 10 Ferrules | 290 MA203 |
N/A
|
|
|||
|
03050252
|
Thermo Scientific™
290MA205 |
MS Interface | Pk. of 5 Nuts | 290 MA205 |
N/A
|
|
|||
|
03050253
|
Thermo Scientific™
290MA207 |
Agilent SSL Injectors | Pk. of 5 Nuts | 290 MA207 |
N/A
|
|
|||
|
03050270
|
Thermo Scientific™
290MT211 |
Thermo Scientific DSQ GC-MS Interface | Pk. of 5 Nuts | 290 MT211 |
N/A
|
|
|||
|
03050271
|
Thermo Scientific™
290MT221 |
Thermo Scientific DSQ GC-MS Interface Kits | Pk. of 10 Ferrules | 290 MT221 |
N/A
|
|
|||
|
03050272
|
Thermo Scientific™
290MT222 |
Thermo Scientific DSQ GC-MS Interface Kits | Pk. of 10 Ferrules | 290 MT222 |
N/A
|
|
|||
|
03050273
|
Thermo Scientific™
290MT223 |
Thermo Scientific DSQ GC-MS Interface Kits | Pk. of 10 Ferrules | 290 MT223 |
N/A
|
|
|||
Description
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
- SilTite metal ferrules and nuts are made from the same material and have the same thermal
- expansion coefficient
- Base of the ferrule is flat and forms a perfect seal with the MS interface
- The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
- Unlike other ferrules, do not need re-tightening after installation
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.