Learn More
Thermo Scientific™ Silanized Glass Vials
GREENER_CHOICE
Save time and minimize waste costs when performing quantitative analysis or storing materials by using these pretreated, silanized vials.
Supplier:  Thermo Scientific™ SAA SV4 2
 View more versions of this product
Catalog No. 12200127
Only null left
Description
Description
Save time and minimize waste costs when performing quantitative analysis or storing materials . Thermo Scientific™ Silanized Glass Vials are treated by vapor phase deposition silanization method. Surface deactivation treatments such as silanization and siliconization may play an important role in preserving the integrity of certain materials or extracts stored in glass containers. Surface modification eliminates active sites on the surfaces of boroslicate glass.
  • Silanized to save time and minimize waste costs when performing quantitative analysis or storing materials
  • Silylating agent reacts with active groups on glass surface so they are less reactive; inhibits materials from adhering onto container surface to maximize recovery of trace analytes
Ordering Information:
Case of 100
Includes:
Certificate of Conformance
Recommended for:
Deactivated glassware
Specifications
Chemical Identifiers
Certificate of Analysis (COA), Certificate of Conformance (COC)
Vial
Specifications
Amber glass vial 15 x 45 closed top closure
0.591 in.
15.01 mm
4 mL
100
4 mL
Screw Cap (Closed-Top)
13-425
45.01 mm
PTFE
Amber
Glass
1.772 in.
0.12 oz.
PTFE
100 Cs.
Certificate of Analysis (COA), Certificate of Conformance (COC)
Vial
Product Suggestions
Product Suggestions