missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Hematoxylin 1

Catalog No. 22050112
Change view
Click to view available options
Quantity:
1 pt.
1.32 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22050112 1 pt.
7221L 1.32 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22050112 Supplier Epredia™ Supplier No. 7221
Edge

Yield visually similar results to Gills 1 and 2 with this uniquely formulated product.

  • No filtration required
  • Stains up to 2500 slides per pint
  • Part of the Signature Series Staining System
  • Crisp, well-defined nuclear detail chromatin delineation

Available in 5-liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging

Specifications

Quantity 1 pt.
Product Type Hematoxylin 1