Learn More
Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Catalog No. 8315
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
Product Type:
Flex 100 Dehydrating Reagent
Flex 80 Dehydrating Reagent
Flex 95 Dehydrating Reagent
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
8315 1.32 gal. Flex 80 Dehydrating Reagent
22046344 1 gal. Flex 100 Dehydrating Reagent
22046346 1 gal. Flex 95 Dehydrating Reagent
8215 1.32 gal. Flex 95 Dehydrating Reagent
22046348 1 gal. Flex 80 Dehydrating Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 8315 Supplier Epredia™ Supplier No. 8315
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
  • Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
  • Excellent for lipid extraction allowing for increased paraffin infiltration
  • May be used as a solvent for special stains
Specifications
Product Type Flex 80 Dehydrating Reagent
Quantity 1.32 gal.