Learn More
Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Catalog No. 22046348
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
Product Type:
Flex 100 Dehydrating Reagent
Flex 80 Dehydrating Reagent
Flex 95 Dehydrating Reagent
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22046348 1 gal. Flex 80 Dehydrating Reagent
22046344 1 gal. Flex 100 Dehydrating Reagent
22046346 1 gal. Flex 95 Dehydrating Reagent
8215 1.32 gal. Flex 95 Dehydrating Reagent
8315 1.32 gal. Flex 80 Dehydrating Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22046348 Supplier Epredia™ Supplier No. 8301R
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
  • Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
  • Excellent for lipid extraction allowing for increased paraffin infiltration
  • May be used as a solvent for special stains
Specifications
Product Type Flex 80 Dehydrating Reagent
Quantity 1 gal.