Learn More
Epredia™ Signature Series™ Clarifier™ 1, 2
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Epredia™ Signature Series Clarifier™ 1, 2.
$115.70 - $595.18
| Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
|
74-02L
|
Epredia™
7402L |
1.32 gal. | Clarifier 2 |
Each for $595.18
|
|
||||
|
22-050-118
|
Epredia™
7441 |
4 x 1 gal. | Clarifier 1 |
Case of 4 Each for $427.71
|
|
||||
|
22-050-119
|
Epredia™
7442 |
4 x 1 gal. | Clarifier 2 |
Case of 4 Each for $320.58
|
|
||||
|
22-050-116
|
Epredia™
SH7401 |
1 gal. | Clarifier 1 |
Each for $163.10
|
|
||||
|
22-050-117
|
Epredia™
SH7402 |
1 gal. | Clarifier 2 |
Each for $115.70
|
|
||||
Description
- Improves contrast between Nuclei and Cytoplasm
- Increases cell transparency
- Removes background staining caused by excessive tissue adhesive
- Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
- Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
- Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2