missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Sign: CONFINED SPACE ENTER BY PERMIT ONLY...OBTAIN AUTHORIZED PERMIT

Catalog No. 19107788
Change view
Click to view available options
Material:
Aluminum
Polyester
Mounting Type:
Corner Holes
Self-adhesive
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Mounting Type
19107788 Polyester Self-adhesive
19107787 Aluminum Corner Holes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19107788 Supplier Brady™ Supplier No. 122649

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve workplace safety with signage. The Brady™ Confined Space Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

  • Abrasion-resistant and chemical-resistant
  • Brady™ signs feature bright colors, bold text, and intuitive pictographs
  • Highly visible and easily understood

Specifications

Finish Laminated
Material Polyester
Quantity 1 Ea.
Description Polyester with polyester overlaminate
Mounting Type Self-adhesive
Height (English) 7 in.
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.01 in.
Width (English) 10 in.
Shape Rectangular
Temperature Range (English) -40°F to 230°F
Languages English
Dimensions (L x W) 10 x 7 in.
Displayed Text CONFINED SPACE ENTER BY PERMIT ONLY PREPARE FOR ENTER BY PERMIT ONLY PREPARE FOR ENTRY IDENTIFY HAZARDS OF PERMIT SPACE. DE-ENERGIZE AND LOCKOUT ALL ENERGY SOURCES. DRAIN, CLEAN AND VENTILATE CONFINED SPACES. ISOLATE CONFINED-DISCONNECT FILL AND DRAIN LINES TEST ATMOSPHERE OXYGEN LEVEL BETWEEN 19.5% AND 23.5%. FLAMMABLE GLASS/VAPORS LESS THAN 10% ID LEL. ALL SUBSTANCES BELOW ESTABLISHED PEL. PREPSARE PERSONAL PROTECTION DEVICES RESPIRATOR, PROTECTIVE CLOTHING, LIFELINE AND SAFETY HAMESS. ATTENDANT AND RESCUE EQUIPENT IN PLACE REVIEW COMMUNICATION REOCEDURES OBTAIN AUTHORIZED PERMIT
Product Type Confined Space Sign
Color Red on White
Show More Show Less