missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Sign BIOHAZARD NO FOOD OR DRINK TO BE STORED IN THIS REFRIGERATOR
Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.
$15.34 - $20.87
Specifications
| Material | Vinyl |
|---|---|
| Mounting Type | Self-adhesive |
| Shape | Rectangular |
| Languages | English |
| Product Type | BioHazard Safety Label |
Description
- Good resistance to most common chemicals, oils and water
- Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
- Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation
- Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
| Vinyl | |
| Rectangular | |
| BioHazard Safety Label |
| Self-adhesive | |
| English | |
| Black on Orange |