missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Sign BIOHAZARD NO FOOD OR DRINK TO BE STORED IN THIS REFRIGERATOR

Catalog No. 17108794
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17108794 10 x 7 in.
17108795 14 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 17108794 missing translation for 'mfr' Brady™ missing translation for 'supplierNo' 126632A

Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.

  • Good resistance to most common chemicals, oils and water
  • Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
  • Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation
  • Service temperature of -40°F to +122°F (-40°C to +50°C)

Specifications

Material Vinyl
Mounting Type Self-adhesive
Shape Rectangular
Languages English
Dimensions (L x W) 10 x 7 in.
Product Type BioHazard Safety Label
Color Black on Orange