missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Sigma-Aldrich™ TLC Plates, PEI-Cellulose Matrix, 100 μm

Catalog No. 111009574
Change view
Click to view available options
Quantity:
25/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111009574 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009574 Supplier MilliporeSigma™ Supelco™ Supplier No. Z12288225EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sigma-Aldrich™ TLC Plates may be to perform thin layer chromatography.

Thin-layer chromatography (TLC) plates for adsorption, partition, and ion-exchange techniques. Six media choices with glass, polyester, and aluminum backings. Silica-based adsorbent layers in Sigma-Aldrich TLC plates incorporate a polymeric binder; highly purified silica gel plates incorporate a gypsum/polymer binder. Silica gel - for weakly to strongly polar separations, pore size 60 Å . High purity silica gel - acid washed for separation of aflatoxins. Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose). Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color. Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60 Å.

Specifications

Backing Material Polyester
Material Polyester
Quantity 25/Pk.
Type TLC Plate
Fluorescent Indicator Yes
For Use With (Application) TLC
Product Line Sigma-Aldrich
Sorbent PEI-Cellulose Matrix
Thickness (Metric) 100 μm
Dimensions (L x W) 20 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm
Show More Show Less

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC