missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Sigma-Aldrich™ Silica Gel, Technical Grade, 230-400 mesh

Catalog No. 111002402
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002402 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002402 Supplier MilliporeSigma™ Supelco™ Supplier No. 7171855KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For preparative liquid chromatography

Silica gel is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.

Specifications

Particle Size 230/400 Mesh
Pore Size 60 Å
Grade Technical Grade
Packaging Poly Bottle
Quantity 5 kg
Surface Area (Metric) ≥480 m2/g (approx.)
Product Line Sigma-Aldrich
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC