missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Sigma-Aldrich™ Silica Gel, Spherical, 40-75 μm
Description
High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.
Specifications
Specifications
| Particle Size | 40 to 75 μm |
| Pore Size | 60 Å |
| Grade | High-purity Grade |
| pH | 7 (5% in H2O, suspension) |
| Quantity | 5 kg |
| Surface Area (Metric) | 450 to 550 m2/g |
| Product Line | Sigma-Aldrich |
| Physical Form | Powder |
| MDL Number | MFCD00011232 |
| CAS | 112926-00-8 |
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC