missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Sigma-Aldrich™ Silica Gel, High-Purity Grade, 35-60 mesh

Catalog No. 111012548
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012548 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012548 Supplier MilliporeSigma™ Supelco™ Supplier No. 6074225KG

For column chromatography

High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.

Specifications

Particle Size 35/60 Mesh
Pore Size 60 Å
Grade High-purity Grade
Quantity 25 kg
Surface Area (Metric) 500 m2/g
Product Line Sigma-Aldrich
Physical Form Powder or Crystals
MDL Number MFCD00011232
CAS 112926-00-8

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC