missing translation for 'onlineSavingsMsg'
Learn More

Sigma Diagnostics™ Eosin Y Solution, Alcoholic with Phloxine

Catalog No. SDHT110316
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDHT110316 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDHT110316 Supplier Sigma Diagnostics™ Supplier No. HT110316500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General-purpose cytoplasmic counterstain

  • Certified Eosin Y, 0.1% (w/v) and certified phloxine B, 0.1% (w/v) in acidified ethanol

Specifications

Color Bright pink to pinkish-orange
Format Solution
For Use With (Application) Used with hematoxylin and eosin staining
Quantity 500 mL
Storage Requirements 18–26°C
Product Type Alcoholic Eosin Y Stain