missing translation for 'onlineSavingsMsg'
Learn More

Sigma Diagnostics™ Phosphate buffer pH 7.2 at 25°C

Catalog No. SDP3288
Encompass
Change view
Click to view available options
Quantity:
12 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDP3288 12 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDP3288 Supplier Sigma Diagnostics™ Supplier No. P328812VL

For use as a buffer in various Romanowsky type staining procedures

  • Reconstitution: Each vial will prepare 3.8L of buffer, 8.3mM.

Specifications

Quantity 12 Vials
Storage Requirements 25°C
Format Powder
Concentration 8.3mM
For Use With (Application) Intended for use as a Buffer in Wright stain, Wright Giemsa, Giemsa, May Grunwald, Jenner and Leishman staining procedures
Color White
pH 7.2
Product Type Phosphate buffer pH 7.2 at 25°C