missing translation for 'onlineSavingsMsg'
Learn More

Sigma Diagnostics™ Giemsa stain, modified

Catalog No. 01288062
Change view
Click to view available options
Quantity:
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01288062 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288062 Supplier Sigma Diagnostics™ Supplier No. GS1L1L*

Used in staining blood or bone marrow films

  • When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration
  • The use of purified eosin and thiazine dyes minimizes lot-to-lot variation

Specifications

Quantity 1L
Storage Requirements 18-26°C
Format Solution
Concentration 0.4% w/v in Buffered Methanol solution
For Use With (Application) Used in staining blood or bone marrow films
Color Faint Blue to Dark Blue
pH 6.8
Packaging 1 L in poly bottle
Synonym Giemsa Solution
Product Type Giemsa stain, modified