missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Sigma-Aldrich™ Flash Silica (Dried, Spherical)

Catalog No. 111010960
Change view
Click to view available options
Includes:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
111010960 100 g
111008788 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111010960 Supplier MilliporeSigma™ Supelco™ Supplier No. 97728U

High-purity grade

  • Pore Size: 0.7-0.9 cm3/g pore volume
  • Surface Area: 480 m2/g
  • MDL Number: MFCD00011232
  • BP: 2230°C
  • MP: gt;1600°C
  • CAS: 112926-00-8

Specifications

Includes 100 g
For Use With (Application) Flash Chromatography
Product Line Sigma-Aldrich

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC