missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple, 0.04% (w/v) Indicator Solution, Spectrum™ Chemical

Catalog No. 18609236
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18609236 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18609236 Supplier Spectrum Chemical Mfg Cor Supplier No. B169500MLPL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62625-30-3 | 540.22 g/mol

Specifications

Chemical Name or Material Bromocresol Purple
CAS 62625-30-3, 7732-18-5, 115-40-2
Concentration 0.04%
Packaging Poly Bottle
Visual Transition Interval pH 5.2 (yellow)
Color Dark Purple
Quantity 500 mL
Molecular Weight (g/mol) 540.22
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.