Learn More
ACROBiosystems Inc ACROBiosystems Biotinylated Human Siglec-10 Protein, Fc,Avitag™
Catalog No. 501956541
Encompass
Recombinant Protein;200UG;Biotinylated Human Siglec-10, Fc,Avitag (SI0-H82F6) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 86.4 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Siglec-10, Fc,Avitag (Cat. No. SI0-H82F6) at 5 g/mL (100 L/well) on streptavidin precoated (0.5 g/well) plate, can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0.6-20 ng/mL (QC tested).
Catalog No. 501956541 Supplier ACROBiosystems Inc Supplier No. SI0H82F6200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;200UG;Biotinylated Human Siglec-10, Fc,Avitag (SI0-H82F6) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 86.4 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Siglec-10, Fc,Avitag (Cat. No. SI0-H82F6) at 5 g/mL (100 L/well) on streptavidin precoated (0.5 g/well) plate, can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0.6-20 ng/mL (QC tested).