missing translation for 'onlineSavingsMsg'
Learn More

Shvo's Catalyst, TCI America™

Catalog No. H1322100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1322100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1322100MG Supplier TCI America Supplier No. H1322100MG
Only null left

Chemical Identifiers

CAS 104439-77-2
Molecular Formula C62H42O6Ru2
MDL Number MFCD03840556
Synonym Shvo′Cs Catalyst

Specifications

Melting Point 227°C
Quantity 100 mg
Formula Weight 1085.15
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxytetraphenylcylclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldir